Learn More
Perkin Elmer US LLC Custom Standard HI 1A, 500 mL
Supplier: Perkin Elmer US LLC N9308341
Al @ 10 µg/mL, Mg @ 20 µg/mL Ca @ 32 µg/mL Cu, Fe, Pb, Zn @ 50 µg/mL B @ 1000 µg/mL, P @2000 µg/mL K @ 5000 µg/mL Na @10,000 µg/mL Matrix: 3% HNO3 (v/v) Volume: 500 mL