Learn More
Chemglass Life Sciences Flask Support, Polypropylene
Supplier: Chemglass Life Sciences CG3056P01
Flask Support, Polypropylene. For use with round bottom flasks up to 10 Liters. The flask stand is ribbed to safely hold flasks in place and is fully autoclavable. Stand is 7 in diameter and 2 high.